Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: P2RX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256796.1 | NP_001243725.1 | 10, 11 | 1338 | ||
NM_001261397.1 | NP_001248326.1 | 9, 10 | 1209 | |||
NM_001261398.1 | NP_001248327.1 | 9, 11 | 1290 | |||
NM_002560.2 | NP_002551.2 | 9, 10 | 1290 | |||
NR_046372.1 | 10, 11 | 1328 | ||||
NR_046373.1 | 8, 9 | 1142 | ||||
XM_011538416.2 | XP_011536718.1 | 5, 6 | 653 | |||
GenBank mRNA | AF000234.1 | - | 9, 10 | 886 | ||
AK225604.1 | - | 9, 11 | 986 | |||
AK293599.1 | - | 6, 7 | 780 | |||
AK293660.1 | - | 8, 9 | 925 | |||
AK297704.1 | - | 8, 9 | 874 | |||
AK301080.1 | - | 9, 10 | 974 | |||
AK308549.1 | - | 10, 11 | 1060 | |||
AK308868.1 | - | 9, 10 | 1981 | |||
BC017458.1 | - | 9, 10 | 1019 | |||
BC033826.1 | - | 9, 10 | 1010 | 5216449 | ||
BT019738.1 | - | 9, 10 | 986 | |||
BT019739.1 | - | 9, 10 | 905 | |||
JF807485.1 | - | 2, 3 | 143 | |||
JX471561.1 | - | 2, 3 | 182 | |||
U83993.1 | - | 9, 10 | 1295 | |||
Y07684.1 | - | 9, 10 | 1013 |