Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nagk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164187.1 | NP_001157659.1 | 6 | 721 | ||
NM_019542.2 | NP_062415.1 | 6 | 730 | |||
GenBank mRNA | AJ242909.1 | - | 6 | 742 | ||
AJ242911.1 | - | 6 | 682 | |||
AK007240.1 | - | 6 | 725 | |||
AK155292.1 | - | 6 | 622 | |||
BC004689.1 | - | 6 | 608 | 3501470 |