Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hao2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019545.4 | NP_062418.3 | 9 | 1451 | ||
GenBank mRNA | AF272947.1 | - | 9 | 1447 | ||
AK018684.1 | - | 9 | 1439 | |||
AK078908.1 | - | 9 | 1441 | |||
AK143991.1 | - | 9 | 1442 | |||
BC027754.1 | - | 9 | 1483 | 5064100 |