Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTK6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256358.1 | NP_001243287.1 | 3, 4 | 599 | ||
NM_005975.3 | NP_005966.1 | 4, 5 | 721 | |||
GenBank mRNA | AK301364.1 | - | 3, 4 | 599 | ||
AK315232.1 | - | 4, 5 | 703 | |||
BC035843.1 | - | 4, 5 | 705 | 5746034 | ||
X78549.1 | - | 4, 5 | 693 |