Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tspan5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019571.5 | NP_062517.1 | 8 | 1828 | ||
GenBank mRNA | AF121344.1 | - | 8 | 1842 | ||
AK013350.1 | - | 8 | 1808 | |||
AK033543.1 | - | 8 | 1527 | |||
AK194230.1 | - | 1 | 171 | |||
AK195043.1 | - | 1 | 171 | |||
AK196262.1 | - | 1 | 171 | |||
AK198383.1 | - | 1 | 171 | |||
AK199840.1 | - | 1 | 174 | |||
AK201050.1 | - | 1 | 172 | |||
AK204282.1 | - | 1 | 171 | |||
BC046784.1 | - | 8 | 1667 | 6416107 | ||
BC058695.1 | - | 8 | 1799 | 6813872 |