Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gtpbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145979.1 | NP_001139451.1 | 8, 9 | 1318 | ||
NM_019581.3 | NP_062527.2 | 8, 9 | 1353 | |||
GenBank mRNA | AB024573.1 | - | 6, 7 | 989 | ||
AF168991.1 | - | 8, 9 | 1258 | |||
AK087511.1 | - | 8, 9 | 1224 | |||
AK146415.1 | - | 8, 9 | 1303 | |||
AK148020.1 | - | 8, 9 | 1275 | |||
BC049089.1 | - | 8, 9 | 1314 | 5720710 | ||
BC051958.1 | - | 8, 9 | 1314 | 5720710 |