Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbc1d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289514.1 | NP_001276443.1 | 5, 6 | 956 | ||
NM_019636.3 | NP_062610.2 | 6, 7 | 1537 | |||
GenBank mRNA | AK122445.1 | - | 6, 7 | 2346 | ||
AK135665.1 | - | 5, 6 | 1127 | |||
AK135892.1 | - | 5, 6 | 1221 | |||
AK148356.1 | - | 5, 6 | 912 | |||
AK154591.1 | - | 7, 8 | 1573 | |||
AK159256.1 | - | 6, 7 | 1533 | |||
AK164923.1 | - | 2, 3 | 241 | |||
BC059824.1 | - | 7, 8 | 1902 | |||
BC117931.1 | - | 5, 6 | 1205 | 40097712 | ||
U33005.1 | - | 6, 7 | 1493 |