Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hsd17b12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019657.4 | NP_062631.1 | 2 | 217 | ||
GenBank mRNA | AF064635.1 | - | 2 | 196 | ||
AK076088.1 | - | 2 | 203 | |||
AK078841.1 | - | 2 | 218 | |||
AK081869.1 | - | 2 | 211 | |||
AK082715.1 | - | 2 | 211 | |||
AK088521.1 | - | 2 | 211 | |||
AK150849.1 | - | 2 | 212 | |||
AK152489.1 | - | 2 | 212 | |||
AK166027.1 | - | 2 | 211 | |||
AK167908.1 | - | 2 | 211 | |||
BC010749.1 | - | 2 | 193 | |||
BC037620.1 | - | 2 | 210 | 5375831 | ||
BC090659.1 | - | 2 | 187 | 30675153 |