Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Net1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019671.3 | NP_062645.2 | 2 | 308 | ||
GenBank mRNA | AF094520.1 | - | 2 | 204 | ||
BY009210.1 | - | 2 | 309 | |||
CF167482.1 | - | 2 | 292 |