Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTK6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256358.1 | NP_001243287.1 | 7 | 1757 | ||
NM_005975.3 | NP_005966.1 | 8 | 1879 | |||
GenBank mRNA | AK124082.1 | - | 3 | 2193 | ||
BC035843.1 | - | 8 | 1863 | 5746034 | ||
X78549.1 | - | 8 | 1851 |