Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNK5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003740.3 | NP_003731.1 | 5 | 1107 | ||
XM_005249456.1 | XP_005249513.1 | 5 | 831 | |||
XM_006715235.2 | XP_006715298.1 | 3 | 370 | |||
GenBank mRNA | AB743589.1 | - | 4 | 576 | ||
AF084830.1 | - | 5 | 1082 | |||
AK314298.1 | - | 5 | 840 | |||
BC060793.1 | - | 5 | 1107 | 30348251 | ||
BC069573.1 | - | 5 | 814 | 7262361 | ||
EU978936.1 | - | 5 | 893 |