Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019727.2 | NP_062701.2 | 3 | 457 | ||
GenBank mRNA | AB019214.1 | - | 3 | 397 | ||
AF154120.1 | - | 3 | 458 | |||
AK149850.1 | - | 3 | 369 | |||
AK154079.1 | - | 3 | 367 | |||
AK155746.1 | - | 3 | 345 | |||
AK161241.1 | - | 3 | 438 | |||
AK167177.1 | - | 3 | 412 | |||
AK167993.1 | - | 3 | 349 | |||
AK168742.1 | - | 3 | 450 | |||
BC066044.1 | - | 3 | 414 | 6807363 | ||
BC066189.1 | - | 3 | 348 | 30471747 |