Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252580.1 | NP_001239509.1 | 3 | 435 | ||
NM_019729.3 | NP_062703.2 | 3 | 402 | |||
GenBank mRNA | AB045709.1 | - | 2 | 176 | ||
AF057146.1 | - | 2 | 193 | |||
AK040791.1 | - | 3 | 349 | |||
AK122195.1 | - | 3 | 313 | |||
AK129003.1 | - | 3 | 403 | |||
AK153015.1 | - | 3 | 313 | |||
AK163469.1 | - | 3 | 335 | |||
AK166540.1 | - | 3 | 403 | |||
BC038123.1 | - | 3 | 387 | |||
BC050947.1 | - | 3 | 367 | 6825981 | ||
BC061465.1 | - | 3 | 367 | 6825981 | ||
BC066126.1 | - | 3 | 365 | 30301665 |