Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acot9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019736.3 | NP_062710.2 | 4 | 411 | ||
GenBank mRNA | AB028898.1 | - | 4 | 315 | ||
AJ238893.1 | - | 4 | 366 | |||
AK002892.1 | - | 4 | 375 | |||
AK009717.1 | - | 4 | 374 | |||
AK076513.1 | - | 5 | 465 | |||
AK088082.1 | - | 4 | 374 | |||
AK168713.1 | - | 4 | 413 | |||
AK170876.1 | - | 4 | 374 | |||
BC021763.1 | - | 4 | 401 | 4458575 |