Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stmn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019641.4 | NP_062615.1 | 3 | 322 | ||
GenBank mRNA | AB064953.1 | - | 3 | 275 | ||
AK002606.1 | - | 3 | 244 | |||
AK012231.1 | - | 3 | 244 | |||
AK012347.1 | - | 3 | 244 | |||
AK013302.1 | - | 3 | 244 | |||
AK014108.1 | - | 3 | 245 | |||
AK088251.1 | - | 3 | 245 | |||
AK088628.1 | - | 3 | 244 | |||
BC010581.1 | - | 3 | 231 | 3154091 | ||
BC031831.1 | - | 3 | 230 | 4500926 | ||
BC054396.1 | - | 2 | 223 | 6492045 | ||
X94915.1 | - | 1 | 152 |
Gene Symbol: Gm11223 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK030082.1 | - | 318 |