Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FKBP1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001322963.1 | NP_001309892.1 | 5 | 776 | ||
NM_001322964.1 | NP_001309893.1 | 4 | 595 | |||
NM_004116.4 | NP_004107.1 | 4 | 708 | |||
NM_054033.3 | NP_473374.1 | 4 | 753 | |||
NR_136536.1 | 6 | 910 | ||||
NR_136538.1 | 5 | 842 | ||||
NR_136539.1 | 4 | 640 | ||||
XM_017003593.1 | XP_016859082.1 | 6 | 811 | |||
XM_017003594.1 | XP_016859083.1 | 4 | 575 | |||
XM_017003595.1 | XP_016859084.1 | 3 | 556 | |||
XM_017003596.1 | XP_016859085.1 | 5 | 802 | |||
GenBank mRNA | BC002614.1 | - | 4 | 696 | 3161896 | |
BU508033.1 | - | 4 | 672 | |||
D38037.1 | - | 4 | 633 | |||
L37086.1 | - | 4 | 625 | |||
S69800.1 | - | 5 | 545 | |||
S69815.1 | - | 4 | 629 |