Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FKBP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145775.2 | NP_001139247.1 | 12 | 2137 | ||
NM_001145776.1 | NP_001139248.1 | 11 | 2024 | |||
NM_004117.3 | NP_004108.1 | 11 | 1979 | |||
GenBank mRNA | AF194172.1 | - | 11 | 1973 | ||
AL122066.1 | - | 1 | 328 | |||
BC042605.1 | - | 12 | 2137 | 4539929 | ||
BC111050.1 | - | 11 | 1940 | 5723416 | ||
BX647390.1 | - | 11 | 1952 | |||
U42031.1 | - | 10 | 1798 |