Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plcb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145830.1 | NP_001139302.1 | 2, 3 | 554 | ||
NM_019677.2 | NP_062651.2 | 2, 3 | 554 | |||
GenBank mRNA | AF022801.1 | - | 1, 2 | 102 | ||
AF498249.1 | - | 2, 3 | 185 | |||
AF498250.1 | - | 2, 3 | 185 | |||
AK040911.1 | - | 2, 3 | 545 | |||
AK141164.1 | - | 2, 3 | 501 | |||
AK141464.1 | - | 2, 3 | 490 | |||
AK143153.1 | - | 2, 3 | 555 | |||
BC058710.1 | - | 2, 3 | 521 | 6842016 | ||
U85712.1 | - | 2, 3 | 188 | |||
U85713.1 | - | 2, 3 | 188 | |||
U85714.1 | - | 2, 3 | 188 |