Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Letm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019694.1 | NP_062668.1 | 3 | 625 | ||
GenBank mRNA | AF061026.1 | - | 3 | 625 | ||
AK133849.1 | - | 3 | 644 | |||
BC023862.1 | - | 3 | 622 | |||
BC046326.1 | - | 3 | 632 | |||
BC055865.1 | - | 3 | 643 | |||
BC061115.1 | - | 3 | 638 | 6533034 | ||
BC061690.1 | - | 3 | 625 |