Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pard6a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047435.2 | NP_001040900.1 | 3 | 522 | ||
NM_001047436.2 | NP_001040901.1 | 3 | 538 | |||
NM_001286344.1 | NP_001273273.1 | 3 | 507 | |||
NM_001286345.1 | NP_001273274.1 | 3 | 435 | |||
NM_019695.3 | NP_062669.2 | 3 | 525 | |||
GenBank mRNA | AF070970.1 | - | 3 | 521 | ||
AK158848.1 | - | 3 | 520 | |||
BC049593.1 | - | 3 | 499 | 6771847 | ||
BC061114.1 | - | 3 | 435 | 6535523 | ||
BG806503.1 | - | 3 | 450 | |||
CK618363.1 | - | 3 | 481 | |||
CX206739.1 | - | 3 | 488 | |||
CX220562.1 | - | 3 | 532 |