Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fads2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019699.1 | NP_062673.1 | 3 | 445 | ||
GenBank mRNA | AF126798.1 | - | 3 | 445 | ||
AK031337.1 | - | 3 | 558 | |||
AK083282.1 | - | 412 | ||||
AK143524.1 | - | 463 | ||||
AK212413.1 | - | 2 | 179 | |||
BC057189.1 | - | 3 | 433 | 5137343 | ||
HQ264057.1 | - | 3 | 338 | |||
HQ264058.1 | - | 3 | 338 |