Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Orc6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019716.2 | NP_062690.2 | 2 | 400 | ||
GenBank mRNA | AF139659.1 | - | 2 | 241 | ||
BC048689.1 | - | 2 | 109 | |||
BC080739.1 | - | 2 | 119 | 30459034 |