Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc22a21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019723.2 | NP_062697.1 | 10 | 1785 | ||
GenBank mRNA | AB018436.1 | - | 10 | 1785 | ||
AK080177.1 | - | 9 | 1988 | |||
AK133431.1 | - | 10 | 1714 | |||
AK161508.1 | - | 10 | 1681 | |||
BC132147.1 | - | 10 | 1748 | 40130424 | ||
BC137908.1 | - | 10 | 1748 | 8860927 |