Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252580.1 | NP_001239509.1 | 3 | 440 | ||
NM_019729.3 | NP_062703.2 | 3 | 407 | |||
GenBank mRNA | AB045709.1 | - | 2 | 181 | ||
AF057146.1 | - | 2 | 198 | |||
AK040791.1 | - | 3 | 354 | |||
AK122195.1 | - | 3 | 318 | |||
AK129003.1 | - | 3 | 408 | |||
AK153015.1 | - | 3 | 318 | |||
AK163469.1 | - | 3 | 340 | |||
AK166540.1 | - | 3 | 408 | |||
BC038123.1 | - | 3 | 392 | |||
BC050947.1 | - | 3 | 372 | 6825981 | ||
BC061465.1 | - | 3 | 372 | 6825981 | ||
BC066126.1 | - | 3 | 370 | 30301665 |