Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nme3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019730.2 | NP_062704.2 | 5 | 564 | ||
GenBank mRNA | AF153449.1 | - | 5 | 576 | ||
AF288689.1 | - | 5 | 558 | |||
AK002525.1 | - | 5 | 544 | |||
AK007399.1 | - | 5 | 564 | |||
BC028503.1 | - | 5 | 555 | 3326648 |