Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stam2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019667.2 | NP_062641.1 | 14 | 1780 | ||
GenBank mRNA | AK004604.2 | - | 14 | 1780 | ||
AK035789.1 | - | 1833 | ||||
AK159699.1 | - | 14 | 1809 | |||
AK168729.1 | - | 14 | 1812 | |||
BC013818.1 | - | 14 | 1816 | 3671426 |