Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ruvbl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019685.2 | NP_062659.1 | 4 | 488 | ||
GenBank mRNA | AF100694.1 | - | 4 | 527 | ||
AK012170.1 | - | 4 | 436 | |||
AK132783.1 | - | 4 | 470 | |||
AK145062.1 | - | 437 | ||||
AK146258.1 | - | 4 | 480 | |||
AK146374.1 | - | 4 | 481 | |||
AK156078.1 | - | 4 | 458 | |||
BC004718.1 | - | 4 | 454 | 3582426 |