Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPIG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004792.2 | NP_004783.2 | 10 | 797 | ||
XM_005246966.1 | XP_005247023.1 | 10 | 754 | |||
XM_005246967.1 | XP_005247024.1 | 10 | 801 | |||
XM_017005302.1 | XP_016860791.1 | 8 | 593 | |||
GenBank mRNA | AK307552.1 | - | 7 | 588 | ||
BC001555.1 | - | 11 | 757 | |||
BC063711.1 | - | 10 | 899 | |||
BC073875.1 | - | 9 | 652 | |||
BC109373.1 | - | 10 | 801 | |||
BC111693.1 | - | 9 | 739 | 40034815 | ||
U40763.1 | - | 11 | 1062 | |||
X99717.1 | - | 10 | 734 |