Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hbs1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042593.1 | NP_001036058.1 | 8 | 1195 | ||
NM_019702.2 | NP_062676.2 | 8 | 1204 | |||
GenBank mRNA | AF087672.1 | - | 5 | 1065 | ||
AK146678.1 | - | 8 | 1161 | |||
AK166140.1 | - | 8 | 1158 | |||
AK168671.1 | - | 8 | 1158 | |||
AK169281.1 | - | 8 | 1158 | |||
AK173091.1 | - | 8 | 1150 | |||
BC010251.1 | - | 8 | 1129 | 3964560 |