Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNK6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004823.1 | NP_004814.1 | 3 | 2574 | ||
GenBank mRNA | AF117708.1 | - | 3 | 2574 | ||
AF134149.1 | - | 3 | 2583 | |||
AK022344.1 | - | 3 | 2303 | |||
BC004367.1 | - | 3 | 2504 | 3639657 |