Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbck1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083921.1 | NP_001077390.1 | 9 | 1521 | ||
NM_019705.3 | NP_062679.2 | 10 | 1362 | |||
GenBank mRNA | AF124663.1 | - | 8 | 1131 | ||
AK085559.1 | - | 9 | 1510 | |||
AK088591.1 | - | 9 | 1483 | |||
AK146176.1 | - | 10 | 1536 | |||
AK166075.1 | - | 9 | 1489 | |||
AK186100.1 | - | 1 | 134 | |||
AK199876.1 | - | 2 | 159 | |||
BC032089.1 | - | 2 | 966 | |||
BC034555.1 | - | 10 | 1358 | 4192310 |