Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcmf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019715.2 | NP_062689.2 | 7 | 2328 | ||
GenBank mRNA | AK142436.1 | - | 7 | 2085 |