Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Orc6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163791.1 | NP_001157263.1 | 4 | 639 | ||
NM_019716.2 | NP_062690.2 | 5 | 769 | |||
GenBank mRNA | AF139659.1 | - | 5 | 610 | ||
AK078405.1 | - | 4 | 373 | |||
AK163256.1 | - | 4 | 583 | |||
BC080739.1 | - | 5 | 488 | 30459034 |