Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mettl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019721.2 | NP_062695.2 | 9 | 1609 | ||
GenBank mRNA | AF135789.1 | - | 7 | 1172 | ||
AK009492.1 | - | 6 | 670 | |||
AK035061.1 | - | 8 | 1427 | |||
AK085189.1 | - | 9 | 1596 | |||
BC012526.1 | - | 9 | 1593 | 3981018 |