Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STK10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005990.3 | NP_005981.3 | 1, 2 | 464 | ||
GenBank mRNA | AB015718.1 | - | 1, 2 | 214 | ||
AK313350.1 | - | 2, 3 | 219 | |||
BC070077.1 | - | 1, 2 | 378 | 5267449 | ||
BI459912.1 | - | 1, 2 | 383 | |||
DA507768.1 | - | 1, 2 | 464 |