Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdcd10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019745.4 | NP_062719.2 | 3, 4 | 412 | ||
GenBank mRNA | AF159368.1 | - | 3, 4 | 315 | ||
AK005594.1 | - | 3, 4 | 341 | |||
AK075926.1 | - | 3, 4 | 360 | |||
AK077308.1 | - | 3, 4 | 389 | |||
AK137198.1 | - | 3, 4 | 382 | |||
AK149182.1 | - | 3, 4 | 318 | |||
AK160234.1 | - | 3, 4 | 362 | |||
AK168857.1 | - | 3, 4 | 346 | |||
AK169689.1 | - | 3, 4 | 346 | |||
BC019650.1 | - | 3, 4 | 352 | 4022513 | ||
BC086778.1 | - | 3, 4 | 331 | 5689088 |