Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptges3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019766.4 | NP_062740.1 | 3 | 438 | ||
GenBank mRNA | AB024935.1 | - | 3 | 246 | ||
AF153479.1 | - | 2 | 135 | |||
AK008805.1 | - | 3 | 469 | |||
AK010636.1 | - | 3 | 440 | |||
AK075932.1 | - | 3 | 303 | |||
AK075987.1 | - | 3 | 349 | |||
AK077538.1 | - | 3 | 439 | |||
AK167443.1 | - | 3 | 414 | |||
AK168073.1 | - | 3 | 349 | |||
AK168210.1 | - | 3 | 349 | |||
AK168721.1 | - | 3 | 288 | |||
AY281130.1 | - | 3 | 465 | |||
BC003708.1 | - | 3 | 331 | 3489418 | ||
BC085264.1 | - | 3 | 335 | 6392001 |