Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCND1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004979.5 | NP_004970.3 | 6 | 3695 | ||
XM_011543910.2 | XP_011542212.1 | 7 | 2603 | |||
XM_017029508.1 | XP_016884997.1 | 7 | 2434 | |||
XM_017029509.1 | XP_016884998.1 | 7 | 2431 | |||
GenBank mRNA | AB021865.1 | - | 6 | 3419 | ||
AF166003.1 | - | 6 | 2172 | |||
AJ005898.1 | - | 6 | 2122 | |||
AK094431.1 | - | 5 | 2281 | |||
BC045659.1 | - | 6 | 3693 | 5271042 |