Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vps29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019780.1 | NP_062754.1 | 2 | 154 | ||
GenBank mRNA | AB578902.1 | - | 3 | 164 | ||
AB578903.1 | - | 3 | 133 | |||
AB578904.1 | - | 2 | 169 | |||
AB578905.1 | - | 2 | 158 | |||
AB578906.1 | - | 2 | 152 | |||
AB578907.1 | - | 2 | 151 | |||
AB578908.1 | - | 2 | 123 | |||
AB578909.1 | - | 2 | 168 | |||
AB578910.1 | - | 2 | 137 | |||
AB578911.1 | - | 2 | 157 | |||
AF193794.1 | - | 2 | 154 | |||
AK004103.1 | - | 3 | 171 | |||
AK008248.1 | - | 3 | 174 | |||
AK036426.1 | - | 2 | 142 | |||
AK150330.1 | - | 2 | 164 | |||
BC005663.1 | - | 2 | 143 | 2645432 |