Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmeff2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019790.4 | NP_062764.1 | 2 | 678 | ||
GenBank mRNA | AB017270.1 | - | 2 | 641 | ||
AB041565.1 | - | 2 | 682 | |||
AK029307.1 | - | 2 | 678 | |||
AK030053.1 | - | 7 | 1914 | |||
AK076254.1 | - | 2 | 678 | |||
AK135038.1 | - | 2 | 665 | |||
BC034850.1 | - | 2 | 654 | 1397175 |