Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnaja2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019794.4 | NP_062768.1 | 1, 2 | 110 | ||
GenBank mRNA | AB028853.1 | - | 1, 2 | 111 | ||
AK031858.1 | - | 1, 2 | 177 | |||
AK077672.1 | - | 1, 2 | 181 | |||
AK083208.1 | - | 1, 2 | 178 | |||
AK163255.1 | - | 1, 2 | 109 | |||
AK169169.1 | - | 1, 2 | 170 | |||
BC003420.1 | - | 1, 2 | 183 | 2582183 |