Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acpp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019807.2 | NP_062781.2 | 1 | 167 | ||
NM_207668.2 | NP_997551.1 | 1 | 167 | |||
GenBank mRNA | AF210243.1 | - | 1 | 120 | ||
AK020693.1 | - | 1 | 128 | |||
AK029273.1 | - | 1 | 153 | |||
AK076383.1 | - | 1 | 174 | |||
AK163330.1 | - | 1 | 154 | |||
BC139826.1 | - | 1 | 158 | 40142642 |