Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Timm22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019818.5 | NP_062792.2 | 1 | 241 | ||
NM_023355.2 | NP_075844.1 | 1 | 241 | |||
GenBank mRNA | AF223950.1 | - | 1 | 207 | ||
AK011745.1 | - | 1 | 193 | |||
AK012130.2 | - | 1 | 195 | |||
AK017389.1 | - | 1 | 194 | |||
AK028132.1 | - | 1 | 193 | |||
AK153837.1 | - | 1 | 196 | |||
BC022610.1 | - | 1 | 177 | |||
BC055279.1 | - | 1 | 205 | 5688754 | ||
BF470824.1 | - | 1 | 217 | |||
BY309364.1 | - | 1 | 243 | |||
CF740208.1 | - | 1 | 201 |