Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Amotl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019764.2 | NP_062738.2 | 4 | 1287 | ||
GenBank mRNA | AF175967.1 | - | 4 | 570 | ||
AF175968.1 | - | 2 | 1427 | |||
AK040912.1 | - | 4 | 1368 | |||
AK077535.1 | - | 4 | 1283 | |||
AK141348.1 | - | 4 | 1287 | |||
AK164279.1 | - | 4 | 1356 | |||
AK164819.1 | - | 4 | 1287 | |||
AK169419.1 | - | 4 | 1287 | |||
AX763686.1 | - | 3 | 1126 | |||
BC027824.1 | - | 4 | 1257 | 5323888 | ||
BC035201.1 | - | 4 | 1269 | |||
BC108411.1 | - | 4 | 1321 |