Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arpc1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019767.2 | NP_062741.1 | 3, 4 | 311 | ||
GenBank mRNA | AB024984.1 | - | 3, 4 | 311 | ||
AK002475.1 | - | 3, 4 | 275 | |||
AK003986.1 | - | 3, 4 | 304 | |||
AK076107.1 | - | 3, 4 | 299 | |||
AK145663.1 | - | 3, 4 | 302 | |||
BC001988.1 | - | 3, 4 | 289 | 3485409 |