Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ikbke | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019777.3 | NP_062751.2 | 4 | 536 | ||
GenBank mRNA | AB016589.1 | - | 2 | 224 | ||
AK088580.1 | - | 4 | 538 | |||
AK149911.1 | - | 4 | 525 | |||
AK153046.1 | - | 4 | 537 | |||
CT010206.1 | - | 2 | 190 |