Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vps29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019780.1 | NP_062754.1 | 2 | 198 | ||
GenBank mRNA | AB578902.1 | - | 3 | 208 | ||
AB578903.1 | - | 3 | 177 | |||
AB578904.1 | - | 2 | 213 | |||
AB578905.1 | - | 2 | 202 | |||
AB578906.1 | - | 2 | 196 | |||
AB578907.1 | - | 2 | 195 | |||
AB578908.1 | - | 2 | 167 | |||
AB578909.1 | - | 2 | 212 | |||
AB578910.1 | - | 2 | 181 | |||
AB578911.1 | - | 2 | 201 | |||
AF193794.1 | - | 2 | 198 | |||
AK004103.1 | - | 3 | 215 | |||
AK008248.1 | - | 3 | 218 | |||
AK036426.1 | - | 2 | 186 | |||
AK044468.1 | - | 1 | 2520 | |||
AK150330.1 | - | 2 | 208 | |||
BC005663.1 | - | 2 | 187 | 2645432 |