Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bloc1s6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019788.3 | NP_062762.1 | 2, 3 | 342 | ||
GenBank mRNA | AK038381.1 | - | 2, 3 | 320 | ||
AK077720.1 | - | 2, 3 | 324 | |||
AK147116.1 | - | 2, 3 | 345 | |||
AK160580.1 | - | 2, 3 | 312 | |||
AK170964.1 | - | 2, 3 | 273 | |||
AK172571.1 | - | 2, 3 | 323 | |||
BC085260.1 | - | 2, 3 | 311 | 6437935 | ||
BY015259.1 | - | 2, 3 | 343 |