Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B4galt4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285793.1 | NP_001272722.1 | 2 | 465 | ||
NM_019804.4 | NP_062778.2 | 2 | 398 | |||
GenBank mRNA | AF142672.1 | - | 2 | 375 | ||
AF158746.1 | - | 2 | 377 | |||
AK018658.1 | - | 2 | 362 | |||
AK041692.1 | - | 2 | 390 | |||
AK045602.1 | - | 2 | 370 | |||
AK050005.1 | - | 2 | 394 | |||
AK052013.1 | - | 2 | 392 | |||
AK053603.1 | - | 2 | 459 | |||
AK085368.1 | - | 2 | 384 | |||
AK136637.1 | - | 2 | 310 | |||
BC013492.1 | - | 2 | 361 | 4218187 | ||
BC031115.1 | - | 2 | 450 | 5026168 |