Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdlim5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB016586.1 | - | 3 | 206 | ||
AB016587.1 | - | 2 | 185 | |||
AB016588.1 | - | 3 | 206 | |||
BC037476.1 | - | 3 | 248 | 4457868 |